Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYCNOS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103497
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103497 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103497 Supplier Invitrogen™ Supplier No. PA5103497
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MYCNOS.

This gene is transcribed in antisense to the v-myc avian myelocytomatosis viral oncogene neuroblastoma derived homolog gene (MYCN). It is thought to encode a small, novel protein that stabilizes MYCN, prevents apoptosis, and promotes cell proliferation. Transcripts at this locus may also act directly as functional RNAs to recruit transcriptional regulators to the promoter of MYCN and stimulate transcription of this oncogene. The MYCNOS gene therefore functions through both RNA and protein products.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYCNOS
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MYCNOS
Gene Accession No. P40205
Gene Alias MYCN opposite strand; MYCN-AS1; MYCNOS; NCYM; N-CYM; NYCM
Gene Symbols MYCNOS
Host Species Rabbit
Immunogen A synthesized peptide derived from human MYCNOS(Accession P40205), corresponding to amino acid residues R73-K109.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10408, 110714
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.