Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mycobacterium avium Antibody (2A4), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10064559AF594
Description
Mycobacterium avium Monoclonal specifically detects Mycobacterium avium in Bacteria samples. It is validated for ELISA.Specifications
Mycobacterium avium | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
Mouse | |
Protein G purified | |
Primary | |
Bacteria | |
IgG1 |
ELISA | |
2A4 | |
ELISA | |
m avium, M. avium | |
Mycobacterium Avium cell extract. | |
0.1 mL | |
Specific for Mycobacterium avium. It does not react with Mycobacterium tuberculosis, Mycobacterium phlei, Corynebacterium bovis or Mycoplasma bovis. | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction