Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mycobacterium avium Monoclonal Antibody (2A4)
GREENER_CHOICE

Catalog No. PIMA183339
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA183339 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA183339 Supplier Invitrogen™ Supplier No. MA183339
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody does not cross-react with Mycobacterium tuberculosis, Mycobacterium phlei, Corynebacterium bovis, or Mycoplasma bovis. Mouse anti Mycobacterium avium, clone 2A4, recognizes Mycobacterium avium. It does not react with Mycobacterium tuberculosis, Mycobacterium phlei, Corynebacterium bovis or Mycoplasma bovis.

Mycobacteria are straight or slightly curved, non motile rods that cause TB. Mycobacteria avium causes TB in birds and a pulmonary infection of humans. It can also be transmitted to immunocompromised humans such as AIDS patients where it can cause disseminated Mycobacterium avium complex (MAC).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mycobacterium avium
Applications ELISA
Classification Monoclonal
Clone 2A4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with <0.1% sodium azide
Gene Alias m avium; M. avium
Host Species Mouse
Immunogen Mycobacterium Avium cell extract.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.