Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mycobacterium avium Monoclonal Antibody (JD11.419.2), Invitrogen™
GREENER_CHOICE

Catalog No. 01671818
Change view
Click to view available options
Quantity:
1 mg
100 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-671-818 500 μg
PIMA546176 100 μg
PIMA546149 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-671-818 Supplier Invitrogen™ Supplier No. MA110788
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-10788 detects Mycobacterim avium. MA1-10788 has been successfully used in ELISA applications. The MA1-10788 immunogen is M. avium cell extract.

Mycobacteria are straight or slightly curved, non motile rods that cause TB. Mycobacteria avium causes TB in birds and a pulmonary infection of humans. It can also be transmitted to immunocompromised humans such as AIDS patients where it can cause disseminated Mycobacterium avium complex (MAC).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mycobacterium avium
Applications ELISA
Classification Monoclonal
Clone JD11.419.2
Concentration 0.9 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Alias m avium; M. avium
Host Species Mouse
Immunogen M. avium cell extract.
Purification Method Protein A
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.