Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mycobacterium avium Monoclonal Antibody (JD12-551.6), Invitrogen™

Catalog No. PIMA546080
Change view
Click to view available options
Quantity:
1 mg
100 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA546080 100 μg
01-671-820 500 μg
PIMA546151 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. PIMA546080 Supplier Invitrogen™ Supplier No. MA546080
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

MA1-10790 detects Mycobacterim avium. MA1-10790 has been successfully used in ELISA applications. The MA1-10790 immunogen is M. avium cell extract.

Mycobacteria are straight or slightly curved, non motile rods that cause TB. Mycobacteria avium causes TB in birds and a pulmonary infection of humans. It can also be transmitted to immunocompromised humans such as AIDS patients where it can cause disseminated Mycobacterium avium complex (MAC).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mycobacterium avium
Applications ELISA
Classification Monoclonal
Clone JD12-551.6
Concentration 1.3 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Alias m avium; M. avium
Host Species Mouse
Immunogen M. avium cell extract.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.