Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mycobacterium tuberculosis Antibody (BGN/1209/3875), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10062769AF594
Description
Mycobacterium tuberculosis Monoclonal specifically detects Mycobacterium tuberculosis in Bacteria samples. It is validated for Western Blot, ELISA.Specifications
Mycobacterium tuberculosis | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
Mouse | |
0.1 mL | |
This product recognises the 38 kDa antigen from Mycobacterium tuberculosis, a peptide believed to be involved in the regulation of proteins necessary for virulence. | |
Store at 4°C in the dark. |
Western Blot, ELISA | |
BGN/1209/3875 | |
Western Blot, ELISA | |
M. tuberculosis | |
Recombinant 38 kDa antigen from Mycobacterium tuberculosis. | |
Primary | |
Bacteria | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction