Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mycobacterium tuberculosis Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$207.00 - $499.50
Specifications
Antigen | Mycobacterium tuberculosis |
---|---|
Concentration | 4.0 mg/mL |
Dilution | Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 |
Applications | Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunohistochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017737
![]() |
Novus Biologicals
NB100656570.1ML |
0.1 mL |
Each for $207.00
|
|
|||||
NB10065657
![]() |
Novus Biologicals
NB10065657 |
0.5 mL |
Each for $499.50
|
|
|||||
Description
Mycobacterium tuberculosis Polyclonal specifically detects Mycobacterium tuberculosis in Bacteria samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Mycobacterium tuberculosis | |
Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
Polyclonal | |
Purified | |
RUO | |
Phosphate buffered saline with 0.1% Sodium Azide | |
Purified PPD from Mycobacterium tuberculosis. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
4.0 mg/mL | |
Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Bacteria | |
M. tuberculosis | |
IgG | |
IgG purified | |
Recognizes PPD from Mycobacterium tuberculosis. It has not been cross absorbed and may react with related micro-organisms. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title