Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mycobacterium tuberculosis CFP-10 Monoclonal Antibody (L36G)
GREENER_CHOICE

Catalog No. PIMA518220
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA518220 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA518220 Supplier Invitrogen™ Supplier No. MA518220
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Purity is >90%.

Tuberculosis is a chronic bacterial infection which remains one of the leading killers of young adults world wide. Mycobacterium tuberculosis enters the alveoli by airborne transmission. They are engulfed by alveolar macrophages but resist destruction. Although the exact mechanism is not known, it is suspected that that Mycrobacterium tuberculosis survives by preventing the fusion of the phagosome and lysosome. As the bacterium accumulates it forms a tubercle, spreads to the lymph nodes and becomes a systemic infection. Although treatments for tuberculosis do exist, new strains of Mycobacterium tuberculosis that appear to have multiple-drug resistance (MDR) pose a considerable threat to public health.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mycobacterium tuberculosis CFP-10
Applications ELISA
Classification Monoclonal
Clone L36G
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias m tuberculosis; M. tuberculosis
Host Species Mouse
Immunogen M. tuberculosis recombinant protein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.