Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mycobacterium Tuberculosis CFP10 Antibody - BSA Free, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NB11058737

Catalog No. NB11058737


Only null left
Add to Cart

Description

Description

Mycobacterium Tuberculosis CFP10 Polyclonal specifically detects Mycobacterium Tuberculosis CFP10 in Bacteria samples. It is validated for Western Blot.
Specifications

Specifications

Mycobacterium Tuberculosis CFP10
Polyclonal
Unconjugated
PBS (pH 7.4) with 15mM Sodium Azide
Rabbit
Antigen Affinity-purified
RUO
The rabbit polyclonal antibody to CFP10 recognizes the CFP10 (Rv3874) protein of Mycobacterium tuberculosis and Mycobacterium bovis.
Store at 4C. Do not freeze.
IgG
Western Blot
1.0 mg/mL
Western Blot 0.5 to 1 μg/mL
10 kDa culture filtrate antigen cfp10, Culture filtrate protein 10, ESAT 6 like protein esxB, esxB, lhp, mtsA10, Secreted antigenic protein MTSA 10, tuberculosis CFP10
CFP10 (Rv3874) of Mycobacterium tuberculosis
0.1 mg
Primary
Bacteria
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only