Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mycobacterium Tuberculosis CFP10 Antibody (KFB16), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP105466B200UL
Description
Mycobacterium Tuberculosis CFP10 Monoclonal specifically detects Mycobacterium Tuberculosis CFP10 in Bacteria samples. It is validated for ELISA, Sandwich ELISA.Specifications
Mycobacterium Tuberculosis CFP10 | |
Monoclonal | |
Biotin | |
PBS | |
Mouse | |
200 μL | |
Bacteria | |
Purified |
ELISA | |
KFB16 | |
ELISA | |
10 kDa culture filtrate antigen cfp10, Culture filtrate protein 10, ESAT 6 like protein esxB, esxB, lhp, mtsA10, Secreted antigenic protein MTSA 10, tuberculosis CFP10 | |
Mycobacterium tuberculosis CFP10 (KFB16) | |
Mycobacterium tuberculosis CFP10 (KFB16) | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction