Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mycobacterium Tuberculosis CFP10 Antibody (KFB16), Novus Biologicals™
SDP

Catalog No. p-7106181 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP105466 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP105466 Supplier Novus Biologicals Supplier No. NBP105466
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mycobacterium Tuberculosis CFP10 Monoclonal specifically detects Mycobacterium Tuberculosis CFP10 in Bacteria samples. It is validated for ELISA, Dot Blot, Sandwich ELISA.

Specifications

Antigen Mycobacterium Tuberculosis CFP10
Applications Dot Blot, ELISA
Classification Monoclonal
Clone KFB16
Conjugate Unconjugated
Dilution Dot Blot, Sandwich ELISA 1:100-1:2000
Gene Alias 10 kDa culture filtrate antigen cfp10, Culture filtrate protein 10, ESAT 6 like protein esxB, esxB, lhp, mtsA10, Secreted antigenic protein MTSA 10, tuberculosis CFP10
Host Species Mouse
Immunogen Mycobacterium tuberculosis CFP10 (KFB16)
Purification Method Protein G purified
Quantity 0.2 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Mycobacterium tuberculosis CFP10 (KFB16)
Target Species Bacteria
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.