Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mycobacterium tuberculosis HSP65 Monoclonal Antibody (J105G)
GREENER_CHOICE

Catalog No. PIMA17421
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA17421 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA17421 Supplier Invitrogen™ Supplier No. MA17421
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The MA1-7421 antibody reacts with Mycobacter. tuberculosis HSP65. Non-reactive with other Mycobacteria. MA1-7421 has been successfully used in ELISA, and Western blot applications. The MA1-7421 antibody was raised against M. tuberculosis Heat shock protein 65.

Tuberculosis is a chronic bacterial infection which remains one of the leading killers of young adults world wide. Mycobacterium tuberculosis enters the alveoli by airborne transmission. They are engulfed by alveolar macrophages but resist destruction. Although the exact mechanism is not known, it is suspected that that Mycrobacterium tuberculosis survives by preventing the fusion of the phagosome and lysosome. As the bacterium accumulates it forms a tubercle, spreads to the lymph nodes and becomes a systemic infection. Although treatments for tuberculosis do exist, new strains of Mycobacterium tuberculosis that appear to have multiple-drug resistance (MDR) pose a considerable threat to public health.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mycobacterium tuberculosis HSP65
Applications ELISA, Western Blot
Classification Monoclonal
Clone J105G
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias m tuberculosis; M. tuberculosis
Host Species Mouse
Immunogen M. tuberculosis Heat shock protein 65.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.