Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mycobacterium Tuberculosis L-Alanine Dehydrogenase Antibody (1C3), Janelia Fluor™ 549, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP105061JF549
Description
Mycobacterium Tuberculosis L-Alanine Dehydrogenase Monoclonal antibody specifically detects Mycobacterium Tuberculosis L-Alanine Dehydrogenase in Bacteria samples. It is validated for Western Blot, ELISASpecifications
Mycobacterium Tuberculosis L-Alanine Dehydrogenase | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Bacteria | |
Purified |
Western Blot, ELISA | |
1C3 | |
Western Blot, ELISA | |
40 kDa antigen, Alanine dehydrogenase, EC 1.4.1.1, MT2850, MTV002.45, Rv2780, TB43 | |
Culture filtrat from Mycobacterium tuberculosis H37Rv | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction