Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Mycobacterium tuberculosis MPT64 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5117508
Description
This antibody has been tested in direct-ELISA.
MPT64 is a protein secreted by the Mycobacterium tuberculosis (Mtb) complex species, not detected in non-tuberculous mycobacteria (NTM) and bacillus Calmette-Guerin strains with RD2 deletion.
Specifications
Mycobacterium tuberculosis MPT64 | |
Polyclonal | |
Unconjugated | |
mpt64 | |
mpt64 | |
Recombinant Mycobacterium tuberculosis Immunogenic protein MPT64 protein (24-228aa). | |
100 μg | |
Primary | |
Bacteria | |
Antibody | |
IgG |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
P9WIN9 | |
Rabbit | |
Protein G | |
RUO | |
885925 | |
-20°C or -80°C if preferred | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction