Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mycobacterium tuberculosis Polyclonal Antibody, FITC
GREENER_CHOICE

Catalog No. PA173134
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA173134 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA173134 Supplier Invitrogen™ Supplier No. PA173134
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The PA1-73134 antibody reacts with Mycobacterium tuberculosis. PA1-73134 has been successfully used in Immunohistochemistry applications. The PA1-73134 antibody was raised against purified PPD from Mycobacterium tuberculosis.

Tuberculosis is a chronic bacterial infection which remains one of the leading killers of young adults world wide. Mycobacterium tuberculosis enters the alveoli by airborne transmission. They are engulfed by alveolar macrophages but resist destruction. Although the exact mechanism is not known, it is suspected that that Mycrobacterium tuberculosis survives by preventing the fusion of the phagosome and lysosome. As the bacterium accumulates it forms a tubercle, spreads to the lymph nodes and becomes a systemic infection. Although treatments for tuberculosis do exist, new strains of Mycobacterium tuberculosis that appear to have multiple-drug resistance (MDR) pose a considerable threat to public health.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mycobacterium tuberculosis
Applications Immunohistochemistry
Classification Polyclonal
Concentration 5 mg/mL
Conjugate FITC
Formulation PBS with 10mg/mL BSA and 0.1% sodium azide; pH 7.2
Gene Alias m tuberculosis; M. tuberculosis
Host Species Rabbit
Immunogen Purified PPD from Mycobacterium tuberculosis.
Purification Method IgG fraction
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.