Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mycobacterium tuberculosis Tb7.7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5122321
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5122321 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5122321 Supplier Invitrogen™ Supplier No. PA5122321
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody reacts with the Mycobacterium tuberculosis protein Tb7.7 (Rv2654c).

Tb7.7 (Rv2654c) Antitoxin component of a type II toxin-antitoxin (TA) system. Upon expression in M.smegmatis neutralizes the effect of cognate toxin Rv2653c.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mycobacterium tuberculosis Tb7.7
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 15 mM sodium azide; pH 7.4
Gene Rv2654c
Gene Accession No. P9WJ11
Gene Symbols Rv2654c
Host Species Rabbit
Immunogen Recombinant Tb7.7 protein (produced in E. coli).
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 888154
Target Species Bacteria
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.