Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mycoplasma bovis Monoclonal Antibody (MY10-151.1), Invitrogen™
GREENER_CHOICE

Catalog No. 01671822
Change view
Click to view available options
Quantity:
1 mg
100 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-671-822 500 μg
PIMA546108 100 μg
PIMA546155 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-671-822 Supplier Invitrogen™ Supplier No. MA110792
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-10792 detects Mycoplasma bovis. MA1-10792 has been successfully used in ELISA applications. The MA1-10792 immunogen is Mycoplasma bovis cells.

This antibody specifically recognize Mycoplasma bovis. It does not cross-react with Mycoplasma genitalium, Mycoplasma californicum, Strep. agalactiae, Staph. aureus or E. coli.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mycoplasma bovis
Applications ELISA
Classification Monoclonal
Clone MY10-151.1
Concentration 1.5 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Alias m bovis; M. bovis; mycoplasmen
Host Species Mouse
Immunogen Mycoplasma bovis cells.
Purification Method Protein A
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.