Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mycoplasma hominis Monoclonal Antibody (6502)
GREENER_CHOICE

Catalog No. PIMA533282
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA533282 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA533282 Supplier Invitrogen™ Supplier No. MA533282
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

No cross reactivity with M. genitalium, M. pneumoniae and U. urealyticum.

Mycoplasma hominis is a species of bacteria that lacks a cell wall around their cell membranes. Mycoplasma hominis is one of the most frequently isolated mycoplasmas from the human genital tract and has been linked to pelvic inflammatory disease and preterm labor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mycoplasma hominis
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone 6502
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias mycoplasmen
Host Species Mouse
Immunogen Inactivated whole cell preparation of Mycoplasma hominis.
Purification Method Protein A
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.