Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mycoplasma hominis p120 Monoclonal Antibody (H37S)
GREENER_CHOICE

Catalog No. PIMA518230
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA518230 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA518230 Supplier Invitrogen™ Supplier No. MA518230
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody does not cross-react with M. genitalium, M. pneumoniae and U. urealyticum. Purity is >90%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Mycoplasma hominis p120
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone H37S
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias M. hominis p120; mycoplasmen
Host Species Mouse
Immunogen Mycoplasma hominis recombinant protein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.