Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mycoplasma pneumoniae Monoclonal Antibody (Q41F)
GREENER_CHOICE

Catalog No. PIMA518235
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA518235 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA518235 Supplier Invitrogen™ Supplier No. MA518235
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody is reactive with 70 and 80 kDa WB bands of M. pneumoniae and does not react with uninfected cells. Purity is >90%.

Although they are the simplest and smallest self-replicating bacteria, Mycoplasma infections tend to be persistent, difficult to detect, diagnose, and cure. Mycoplasmas are surface parasites of human respiratory and urogenital tracts. Mycoplasma pneumoniae is often the causative agent of community-acquired pneumonia. The organism's affinity for respiratory epithelial cells and its ability to secrete hydrogen peroxide are a potent combination, leading to oxidative tissue damage.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mycoplasma pneumoniae
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone Q41F
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias Bacterial pneumonia; M. pneumoniae; mycoplasmen
Host Species Mouse
Immunogen Mycoplasma pneumoniae native protein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.