Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mycoplasma pneumoniae MPN-083 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117544
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117544 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117544 Supplier Invitrogen™ Supplier No. PA5117544
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody has been tested in direct-ELISA.

MPN_083 is an uncharacterized lipoprotein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mycoplasma pneumoniae MPN-083
Applications Western Blot
Classification Polyclonal
Concentration 3.79 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Alias MPN_083; Uncharacterized lipoprotein MPN_083
Host Species Rabbit
Immunogen Recombinant Mycoplasma pneumoniae Uncharacterized lipoprotein MPN_083 protein (22-242aa).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.