Learn More
Invitrogen™ Mycoplasma pneumoniae P1 Monoclonal Antibody (6533)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA547571
Description
Specificity: Specific for the P1 adhesin of M. pneumoniae. Does not cross react with: Ureaplasma urealyticum, Mycoplasma hominis or Mycoplasma genitalium. Best ELISA pairs (capture/conjugate): Mycoplasma pneumoniae P1 Monoclonal antibody (Product # MA5-47571)/Mycoplasma pneumoniae Polyclonal antibody (Product # MA5-33285); Mycoplasma pneumoniae P1 Monoclonal antibody (Product # MA5-47571)/Mycoplasma pneumoniae P1 Monoclonal antibody (Product # MA5-47573). Purified preparations consist of >90% pure mouse monoclonal antibody which has been purified from ascites fluid by protein A chromatography. The final preparation is formulated to a protein concentration of 100 μg/mL in 0.01 M phosphate buffered saline, pH 7.2 and contains 0.1% sodium azide. Each vial contains 1.0 mL. This product contains no stabilizing proteins and should be stored at 4C until ready for use. Working dilution must be determined by the user. Suggested starting ranges are 1:10-1:50 for ICC/IF and 1:20-1:200. for ELISA.
Although they are the simplest and smallest self replicating bacteria, Mycoplasma infections tend to be persistent, difficult to detect, diagnose, and cure. Mycoplasmas are surface parasites of human respiratory and urogenital tracts. Mycoplasma pneumoniae is often the causative agent of community-acquired pneumonia. The affinity of the organism for respiratory epithelial cells and its ability to secrete hydrogen peroxide are a potent combination, leading to oxidative tissue damage.
Specifications
Mycoplasma pneumoniae P1 | |
Monoclonal | |
100 μg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Mouse | |
Protein A | |
RUO | |
Bacteria | |
Antibody | |
IgG1 |
ELISA, Immunocytochemistry | |
6533 | |
Unconjugated | |
M. pneumoniae | |
Partially purified preparation of native Mycoplasma pneumoniae ATCC 15531. | |
100 μg | |
Primary | |
4°C | |
Liquid |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.