Learn More
Invitrogen™ MYCT1 Polyclonal Antibody

Description
MYCT1 Polyclonal Antibody for Western Blot, Flow
Specifications
Specifications
| Antigen | MYCT1 |
| Applications | Flow Cytometry, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | Myct1 |
| Gene Accession No. | Q8N699 |
| Gene Alias | 1110020B04Rik; AI225941; AI642973; MTLC; MTMC1; MT-MC1; myc target 1; myc target in myeloid cells 1; myc target in myeloid cells protein 1; Myc target protein 1; Myct1 |
| Gene Symbols | Myct1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.