Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MYCT1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109999
Description
Immunogen sequence: ASAPISQWSS SRRSRSSYTH GLNRTGFYRH SGCERRSNLS LASLTFQRQA SLEQANSFPR KSSFRASTFH PFLQCPPLPV ETESQLVTLP SSNISPTIST SHSLSRPDYW SSNSLRVGLS TPPPPAYESI IKAFPDS.
MYCT1 was initially identified as a novel target of the c-Myc oncogene in myeloid cells. It is a widely expressed nuclear protein whose overexpression can promote apoptosis, alteration of morphology, enhancement of anchorage-independent cell growth, tumorigenic conversion, promotion of genomic instability, and inhibition of hematopoietic differentiation. MYCT1 binds to the promoters of several c-Myc-regulated genes and it has been suggested that the phenotypes seen in MYCT1-overexpressing cells are a result of the deregulation of these genes. RUNX1-ETO, a fusion protein made up of RUNX1 and ETO, is thought to deregulate the proliferation and responsiveness of human hematopoietic progenitor cells downstream of MYCT1.
Specifications
MYCT1 | |
Polyclonal | |
Unconjugated | |
Myct1 | |
1110020B04Rik; AI225941; AI642973; MTLC; MTMC1; MT-MC1; myc target 1; myc target in myeloid cells 1; myc target in myeloid cells protein 1; Myc target protein 1; Myct1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
292264, 68632, 80177 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
0.51 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q8N699, Q8R411 | |
Myct1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 99-235 of human MYCT1 (NP_079383.2). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction