Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MYCT1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP309497100UL
Description
MYCT1 Polyclonal specifically detects MYCT1 in Human samples. It is validated for Western Blot.Specifications
MYCT1 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
FLJ21269, MGC156309, MGC156310, MTLC, MTLCmyc target in myeloid cells 1, MTMC1, myc target 1, Myc target in myeloid cells protein 1, myc target protein 1 | |
The immunogen is a synthetic peptide directed towards the middle region of Human MYCT1 (NP_079383). Peptide sequence RRRRASAPISQWSSSRRSRSSYTHGLNRTGFYRHSGCERRSNLSLASLTF | |
100 μg | |
Primary | |
Human | |
Purified |
Western Blot | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
80177 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction