Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MyD88 Antibody (4D6), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP227369MFV610
Description
MyD88 Monoclonal antibody specifically detects MyD88 in Human, Mouse, Primate, Macaque samples. It is validated for Flow Cytometry, Flow (Intracellular)Specifications
MyD88 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse, Primate, Macaque | |
Purified |
Flow Cytometry | |
4D6 | |
Flow Cytometry, Flow (Intracellular) | |
MYD88D, myeloid differentiation primary response gene (88), myeloid differentiation primary response protein MyD88 | |
A portion of amino acids 50-100 of human MyD88 was used as the immunogen for this antibody. | |
0.1 mL | |
Cytokine Research, Immunology, Innate Immunity, Protein Kinase | |
4615 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction