Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MyD88 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP219448
Description
MyD88 Polyclonal specifically detects MyD88 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
MyD88 | |
Polyclonal | |
Unconjugated | |
0.1M Tris-Glycine (pH7), 20% Glycerol with 0.01% Thimerosal | |
MYD88D, myeloid differentiation primary response gene (88), myeloid differentiation primary response protein MyD88 | |
Rabbit | |
33 kDa | |
0.1 mL | |
Cytokine Research, Immunology, Innate Immunity, Protein Kinase | |
4615 | |
Human, Mouse | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
0.93mg/mL | |
Western Blot 1:1000-1:10000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000, Immunohistochemistry-Frozen | |
Q99836 | |
MYD88 | |
Recombinant protein encompassing a sequence within the center region of human MYD88. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse. Species reactivity based on sequence identity: Mouse (82%), Monkey (99%), Pig (86%), Rat (81%), Bovine (84%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction