Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ MyD88 Inhibitor Peptide Set
SDP

Catalog No. NBP2293285 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
2 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP2293285 5 mg
NBP229328 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP2293285 Supplier Novus Biologicals™ Supplier No. NBP2293285MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Applications: Flow Cytometry, ELISA, Immunohistochemistry, In vitro assay, In vivo assay, Functional (Inhibition), Binding Inhibition, Block/Neutralize

Specifications

Host Species Human, Mouse, Rat, Xenopus, Zebrafish
Components 2 x 1mg: MyD88 Homodimerization Inhibitor peptide (lyophilized) DRQIKIWFQNRRMKWKKRDVLPGT (MyD88 homodimerization sequence: RDVLPGT). Molecular weight: 3100
2 x 1mg: Antennapedia Control peptide (lyophilized) DRQIKIWFQNRRMKWKK. Molecular weight: 2361
5mg size will contain 5mg of each peptide (MyD88 Homodimerization Inhibitor peptide and Antennapedia Control peptide)
For Use With (Application) Blocking/Neutralizing, ELISA, Functional (Inhibition), In vitro assay, In vivo assay
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Quantity 5 mg
Product Type MyD88 Inhibitor Peptide Set
Molecular Weight (g/mol) 3100
Inhibitors MyD88
Form Lyophilized

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.