Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYD88 Monoclonal Antibody (2E10D3), Proteintech
SDP

Catalog No. 502333113 Shop All Proteintech Group Inc Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-233-3113 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-233-3113 Supplier Proteintech Group Inc Supplier No. 679691IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MYD88 Monoclonal antibody specifically detects MYD88 in Human, Mouse, Pig, Rabbit, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen MYD88
Applications Western Blot
Classification Monoclonal
Clone 2E10D3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Accession No. Q99836
Gene Alias MYD88; MYD88D
Host Species Mouse
Purification Method Protein A
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4615
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.