Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MYD88 Mouse anti-Human, Clone: OTI1B4, liquid, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene TA502104S

Description
This gene encodes a cytosolic adapter protein that plays a central role in the innate and adaptive immune response. This protein functions as an essential signal transducer in the interleukin-1 and Toll-like receptor signaling pathways. These pathways regulate that activation of numerous proinflammatory genes. The encoded protein consists of an N-terminal death domain and a C-terminal Toll-interleukin1 receptor domain. Patients with defects in this gene have an increased susceptibility to pyogenic bacterial infections. Alternate splicing results in multiple transcript variants.Specifications
MYD88 | |
Monoclonal | |
Unconjugated | |
MYD88 | |
MYD88D | |
Mouse | |
Affinity Chromatography | |
RUO | |
4615 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI1B4 | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q99836 | |
MYD88 | |
Full length human recombit protein of human MYD88 produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction