Learn More
MYD88 Mouse anti-Human, Rat, Clone: OTI1B8, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF502118

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a cytosolic adapter protein that plays a central role in the innate and adaptive immune response. This protein functions as an essential signal transducer in the interleukin-1 and Toll-like receptor signaling pathways. These pathways regulate that activation of numerous proinflammatory genes. The encoded protein consists of an N-terminal death domain and a C-terminal Toll-interleukin1 receptor domain. Patients with defects in this gene have an increased susceptibility to pyogenic bacterial infections. Alternate splicing results in multiple transcript variants.Specifications
MYD88 | |
Monoclonal | |
Unconjugated | |
MYD88 | |
MYD88D | |
Mouse | |
Affinity Chromatography | |
RUO | |
301059, 4615 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI1B8 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q6Y1S1, Q99836 | |
MYD88 | |
Full length human recombit protein of human MYD88 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human, Rat | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.