Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MYD88 Rabbit anti-Human, Clone:RM306, Abnova™
Rabbit monoclonal antibody raised against huamn MYD88.
Supplier: Abnova Corporation MAB16933
Specifications
MYD88 | |
Monoclonal | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100-250) Western Blot (1:100-200) The optimal working dilution should be determined by the end user. |
|
MYD88 | |
Rabbit | |
100 μL | |
This antibody reacts to human myeloid differentiation primary response protein MyD88. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
RM306 | |
myeloid differentiation primary response gene (88) | |
In PBS (50% glycerol, 1% BSA, 0.09% sodium azide) | |
MYD88D | |
A peptide corresponding to N-terminus of human MYD88. | |
4615 | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction