Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MYEF2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA590739
Description
Immunogen sequence: MADANKAEVP GATGGDSPHL QPAEPPGEPR REPHPAEAEK QQPQHSSSSN GVKMENDESA KEEKSDLKEK STGSKKANRF HPYSKDKNSG TGEKKGPNRN RVFISNIPYD MKWQAIKDLM; Positive Samples: Jurkat, Mouse lung, Mouse testis, Mouse brain; Cellular Location: Nucleus.
MEF-2 is expressed early in the differentiation program and is suppressed by specific polypeptide growth factors. The ability of MEF-2 to recognize conserved activating elements associated with multiple-specific genes suggests that this factor may participate in the coordinate regulation of genes during myogenesis.
Specifications
MYEF2 | |
Polyclonal | |
Unconjugated | |
MYEF2 | |
9430071B01; HsT18564; hypothetical protein LOC641483; Kiaa1341; LOW QUALITY PROTEIN: myelin expression factor 2; Mef2; MEF-2; mKIAA1341; MST156; MSTP156; Myef2; myEF-2; myelin basic protein expression factor 2, repressor; myelin expression factor 2; myelin gene expression factor 2; zgc:123351; zgc:152722 | |
Rabbit | |
Affinity chromatography | |
RUO | |
17876, 50804, 679712 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.95 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q8C854, Q9P2K5 | |
MYEF2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-120 of human MYEF2 (NP_0012881391). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction