Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYEF2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540589
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540589 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540589 Supplier Invitrogen™ Supplier No. PA540589
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: QAGRLGSTIF VANLDFKVGW KKLKEVFSIA GTVKRADIKE DKDGKSRGMG Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 85%.

MEF-2 is expressed early in the differentiation program and is suppressed by specific polypeptide growth factors. The ability of MEF-2 to recognize conserved activating elements associated with multiple-specific genes suggests that this factor may participate in the coordinate regulation of genes during myogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYEF2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MYEF2
Gene Accession No. Q8C854, Q9P2K5
Gene Alias 9430071B01; HsT18564; hypothetical protein LOC641483; Kiaa1341; LOW QUALITY PROTEIN: myelin expression factor 2; Mef2; MEF-2; mKIAA1341; MST156; MSTP156; Myef2; myEF-2; myelin basic protein expression factor 2, repressor; myelin expression factor 2; myelin gene expression factor 2; zgc:123351; zgc:152722
Gene Symbols MYEF2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human MYEF2.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17876, 50804
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.