Learn More
MYEF2 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303679AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
MEF-2 is expressed early in the differentiation program and is suppressed by specific polypeptide growth factors. The ability of MEF-2 to recognize conserved activating elements associated with multiple-specific genes suggests that this factor may participate in the coordinate regulation of genes during myogenesis.Specifications
MYEF2 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q9P2K5 | |
MYEF2 | |
Between 1 and 25 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
MYEF2 | |
Myelin expression factor 2, MEF-2, MST156, MSTP156, HsT18564 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
50804 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.