Learn More
MYEF2 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303680A

Description
The recommended shelf life for this product is 1 year from date of receipt.
MEF-2 is expressed early in the differentiation program and is suppressed by specific polypeptide growth factors. The ability of MEF-2 to recognize conserved activating elements associated with multiple-specific genes suggests that this factor may participate in the coordinate regulation of genes during myogenesis.Specifications
MYEF2 | |
Polyclonal | |
Unconjugated | |
MYEF2 | |
Myelin expression factor 2, MEF-2, MST156, MSTP156, HsT18564 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
50804 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q9P2K5 | |
MYEF2 | |
Between 25 and 50 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.