Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myelin Basic Protein Ab-1 Rabbit Polyclonal Antibody, Epredia™

Catalog No. RB1460A Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 mL
Form:
Concentrated
Prediluted
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
RB1460A 1 mL Concentrated
RB1460R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RB1460A Supplier Epredia Supplier No. RB1460A

Myelin Basic Protein Ab-1, Rabbit Polyclonal Antibody is one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available.

Myelin basic protein accounts for about 30% of the proteins in myelin from the central nervous system. It is best known for its capacity to induce experimental allergic encephalomyelitis (EAE), a T-lymphocyte mediated disease due to delayed-type hypersensitivity; though each animal species appears to respond to a different fragment of the 170 amino acid polypeptide. Myelin basic protein is expressed in oligodendrocytes, myelin of white matter in the brain and spinal cord and in peripheral nerves.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human. Others not known.

Immunogen: Myelin basic protein isolated from human brain

Positive Control: Brain

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin )

Specifications

Antigen Myelin Basic Protein Ab-1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
Immunogen Myelin basic protein isolated from human brain
Quantity 1 mL
Regulatory Status IVD
Research Discipline Neuroscience
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.