Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myelin Basic Protein Rabbit anti-Human, Rat, Unconjugated, Polyclonal, MilliporeSigma™

Catalog No. ABN91225UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN91225UG 25 μg
ABN912-100UG 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ABN91225UG Supplier MilliporeSigma Supplier No. ABN91225UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Myelin Basic Protein Antibody, Cat. No. ABN912, is a highly specific rabbit polyclonal antibody that targets Myelin basic protein and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Myelin Basic Protein
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias MBP
Gene Symbols MBP
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to 19 amino acids from the internal region of Guinea pig myelin basic protein.
Molecular Weight of Antigen ∽17 kDa observed; 18.21 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Test Specificity This rabbit polyclonal antibody detects myelin basic protein. It targets an epitope within 19 amino acids from the internal region.
Target Species Human, Rat
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.