Learn More
Myelin basic protein Recombinant Rabbit Monoclonal Antibody (ZR109), RAbMono™, Zeta
Rabbit Recombinant Monoclonal Antibody Cocktail
Supplier: ZETA CORPORATION Z2411RS

Description
A recommended positive control tissue for this product is Brain white master, however positive controls are not limited to this tissue type. The primary antibody is intended for laboratory professional use in the detection of the corresponding protein in formalin-fixed, paraffin-embedded tissue stained in manual qualitative immunohistochemistry (IHC) testing. This antibody is intended to be used after the primary diagnosis of tumor has been made by conventional histopathology using non-immunological histochemical stains.
Myelin basic protein Recombinant Monoclonal antibody specifically detects Myelin basic protein in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Myelin basic protein | |
Monoclonal | |
100 μg/mL | |
tris with BSA, NP-40 and <0.1% sodium azide | |
P02686 | |
Rabbit | |
Protein A | |
RUO | |
4155 | |
2-8°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
ZR109 | |
Unconjugated | |
MBP | |
20 kDa microtubule-stabilizing protein; C76307; golli mbp; Golli-mbp; Golli-MBP; myelin basic protein; Golli-Mbp; myelin basic protein; myelin basic protein S; Hmbpr; MBP; MBP S; Mbps; MGC99675; microtubule-stabilizing protein; mLd; MOBP; Myelin A1 protein; myelin basic protein; myelin basic protein Golli-mbp; myelin basic protein S; myelin deficient; myelin membrane encephalitogenic protein; Myelin P1 protein; myelin-associated oligodendrocyte basic protein; R75289; shi; shiverer; Unknown (protein for IMAGE:7984318); unnamed protein product | |
Recombinant fragment (around aa 150-250) of human MBP | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.