Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Myelin expression factor 2 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP212765JF646
Description
Myelin expression factor 2 Polyclonal antibody specifically detects Myelin expression factor 2 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
Myelin expression factor 2 | |
Polyclonal | |
Western Blot, Immunoprecipitation | |
FLJ11213, HsT18564, KIAA1341MSTP156, MEF-2myEF-2, MGC87325, MST156, MyEF-2, myelin expression factor 2, myelin gene expression factor 2 | |
The immunogen this antibody was made to, maps to a region between residue 25 and 50 of human Myelin Expression Factor 2 using the numbering given in entry NP_057216.2 (GeneID 50804). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Immunoprecipitation | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
50804 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction