Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Myelin Monoclonal Antibody (2B5)
GREENER_CHOICE

Catalog No. 01672018
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-672-018 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-672-018 Supplier Invitrogen™ Supplier No. MA135712
  • $639.00
    $505.00 / Each of 1

    Save $134.00

    21% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

In paraffin sections antibody pm 43 recognizes only myelin in the peripheral nervous system (PNS). In frozen sections it also reacts with myelin in the central nervous system (CNS). Is useful for studying myelination and demyelination processes in the PNS, and remyelination processes in the CNS as can be observed after trauma in multiple sclerosis. Reacts with a 43 kD protein in immunoblotting. Positive control: Peripheral nerve.

Myelin is a membrane characteristic of the nervous tissue and functions as an insulator to increase the velocity of the stimuli being transmitted between a nerve cell body and its target. Myelin isolated from human and bovine nervous tissue is composed of approximately 80% lipid and 20% protein, and 30% of the protein fraction constitutes myelin basic protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Myelin
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 2B5
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with 0.09% sodium azide
Gene Alias BTN6; BTNL11; MGC99675; MOG; MOG alpha-5; MOG AluA; MOG AluB; MOG Ig-AluB; MOGIG2; myelin oligodendrocyte glycoprotein; myelin-oligodendrocyte glycoprotein; NRCLP7
Host Species Mouse
Immunogen Human myelin isolated from peripheral nervous system.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mammalia, Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.