Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Myelin PLP Antibody (PLP1/4259), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308534AF647
Description
Myelin PLP Monoclonal antibody specifically detects Myelin PLP in Human, Mouse, Rat, Bovine, Reptile samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Myelin PLP | |
Monoclonal | |
Alexa Fluor 647 | |
HLD1, lipophilin, major myelin proteolipid protein, MMPL, myelin proteolipid protein, PLP, PLP/DM20, PMD, proteolipid protein 1, spastic paraplegia 2, uncomplicated, SPG2 | |
Synthetic peptide GRGTKF that recognizes the C terminal region (aa272-277) of Myelin PLP protein. (Uniprot: P60201) | |
100 μg | |
Immune System Diseases, Immunology, Neuroscience, Stem Cell Markers | |
5354 | |
Store at 4°C in the dark. | |
IgG2a |
Immunohistochemistry (Paraffin) | |
PLP1/4259 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Bovine, Reptile | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction