Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Myeloid-Associated Differentiation Marker Antibody (MYADM/971), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311489V
Description
Myeloid-Associated Differentiation Marker Monoclonal antibody specifically detects Myeloid-Associated Differentiation Marker in Human, Macaque samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
Myeloid-Associated Differentiation Marker | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
91663 | |
Store at 4°C in the dark. | |
IgG1 κ |
Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
MYADM/971 | |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
MYADM, Myeloid Upregulated Protein, Protein SB135, SB135 | |
Recombinant full-length human Myeloid-Associated Differentiation Marker protein. (Uniprot: Q96S97) | |
0.1 mL | |
Primary | |
Human, Macaque | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction