Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Myeloid-Associated Differentiation Marker Antibody (MYADM/971), Novus Biologicals™

Mouse Monoclonal Antibody
$268.00 - $526.00
Specifications
| Antigen | Myeloid-Associated Differentiation Marker |
|---|---|
| Clone | MYADM/971 |
| Concentration | 0.2mg/mL |
| Dilution | Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, SDS-Page, Immunofluorescence 1 - 2 ug/ml |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), SDS-Page, Immunofluorescence |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP24435800
![]() |
Novus Biologicals
NBP2443580.02MG |
0.02 mg |
Each for $268.00
|
|
|||||
NBP24435801
![]() |
Novus Biologicals
NBP2443580.1MG |
0.1 mg |
Each for $526.00
|
|
|||||
NBP24435802
![]() |
Novus Biologicals
NBP2443580.2MG |
0.2 mg | N/A | N/A | N/A | ||||
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence
Myeloid-Associated Differentiation Marker Monoclonal specifically detects Myeloid-Associated Differentiation Marker in Human, Monkey, Rhesus Macaque samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence.Specifications
| Myeloid-Associated Differentiation Marker | |
| 0.2mg/mL | |
| Flow Cytometry, Immunohistochemistry (Paraffin), SDS-Page, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Human, Monkey | |
| MYADM, Myeloid Upregulated Protein, Protein SB135, SB135 | |
| MYADM | |
| IgG1 | |
| Protein A or G purified | |
| Recognizes a myeloid associated differentiation antigen in the cytoplasm of mature granulocytes. It shows no reactivity with any other cell type in human tissues. Markers of myeloid cells are useful in the identification of different levels of cellular differentiation. It reacts with early precursor and mature forms of human and monkey myeloid cells. This MAb is useful for the detection of myeloid leukemias and granulocytic sarcomas. It can be used as a marker of granulocytes in normal tissues or inflammatory processes. |
| MYADM/971 | |
| Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, SDS-Page, Immunofluorescence 1 - 2 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
| 91663 | |
| Recombinant human MYADM protein | |
| Primary | |
| Store at 4C. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title