Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Myeloid-Associated Differentiation Marker Antibody (MYADM/972), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311468PCP
Description
Myeloid-Associated Differentiation Marker Monoclonal antibody specifically detects Myeloid-Associated Differentiation Marker in Human, Macaque samples. It is validated for Flow CytometrySpecifications
Myeloid-Associated Differentiation Marker | |
Monoclonal | |
PerCP | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
91663 | |
Store at 4°C in the dark. | |
IgG1 κ |
Flow Cytometry | |
MYADM/972 | |
Flow Cytometry | |
MYADM, Myeloid Upregulated Protein, Protein SB135, SB135 | |
Recombinant human Myeloid-Associated Differentiation Marker protein (Uniprot: Q96S97) | |
0.1 mL | |
Primary | |
Human, Macaque | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction