Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Myeloid Cell Marker Antibody (BM-2), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314086JF646
Description
Myeloid Cell Marker Monoclonal antibody specifically detects Myeloid Cell Marker in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| Myeloid Cell Marker | |
| Monoclonal | |
| Janelia Fluor 646 | |
| myeloid specific antigen, myeloid specific protein, p183 DNA-binding cytoplasmic ptotein | |
| Nuclei from pokeweed mitogen stimulated human peripheral blood lymphocytes | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| BM-2 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction