Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Myeloid leukemia factor 1 Antibody (OTI3F11), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272849FR
Description
Myeloid leukemia factor 1 Monoclonal specifically detects Myeloid leukemia factor 1 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Myeloid leukemia factor 1 | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
MLF1 | |
Full length recombinant protein of human Myeloid leukemia factor 1 (NP_071888) produced in E.coli. | |
0.1 mL | |
Primary | |
Human | |
IgG2b |
Western Blot, Immunohistochemistry | |
OTI3F11 | |
Western Blot, Immunohistochemistry | |
Myelodysplasia-myeloid leukemia factor 1, myeloid leukemia factor 1 | |
Mouse | |
Affinity Purified | |
Cancer, Stem Cell Markers | |
4291.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction