Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Myeloid Lineage Monoclonal Antibody (OX-82), Biotin
GREENER_CHOICE

Catalog No. PIMA517560
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA517560 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA517560 Supplier Invitrogen™ Supplier No. MA517560
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-17560 targets Myeloid Lineage in FACS, IHC (F), and WB applications and shows reactivity with Rat samples. The MA5-17560 immunogen is anemic rat bone marrow.

The myeloid lineage encompasses a diverse range of hematopoietic cells derived from the common myeloid progenitor (CMP) in the bone marrow.These cells include granulocytes (neutrophils, eosinophils, basophils), monocytes, macrophages, dendritic cells, erythrocytes, megakaryocytes, and mast cells. Myeloid cells play pivotal roles in innate immunity, inflammation, and tissue homeostasis. The differentiation and functional states of myeloid cells are regulated by various growth factors and cytokines, revealing significant heterogeneity within tissues. Dysregulation or aberrant differentiation of myeloid cells can contribute to pathological conditions, including autoimmune diseases and cancer, emphasizing the clinical importance of understanding myeloid cell biology.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Myeloid Lineage
Applications Flow Cytometry, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone OX-82
Concentration 0.1 mg/mL
Conjugate Biotin
Formulation PBS with 4-5mg/mL BSA and 0.02% sodium azide
Host Species Mouse
Immunogen Anemic rat bone marrow.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.