Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Myeloid Lineage Monoclonal Antibody (OX-82)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA170034
Description
MA1-70034 detects Myeloid Lineage in rat samples. MA1-70034 has been successfully used in flow cytometry, western blot, immunohistochemistry (frozen sections) procedures. The MA1-70034 immunogen is anemic rat bone marrow.
The myeloid lineage encompasses a diverse range of hematopoietic cells derived from the common myeloid progenitor (CMP) in the bone marrow.These cells include granulocytes (neutrophils, eosinophils, basophils), monocytes, macrophages, dendritic cells, erythrocytes, megakaryocytes, and mast cells. Myeloid cells play pivotal roles in innate immunity, inflammation, and tissue homeostasis. The differentiation and functional states of myeloid cells are regulated by various growth factors and cytokines, revealing significant heterogeneity within tissues. Dysregulation or aberrant differentiation of myeloid cells can contribute to pathological conditions, including autoimmune diseases and cancer, emphasizing the clinical importance of understanding myeloid cell biology.
Specifications
Myeloid Lineage | |
Monoclonal | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Anemic rat bone marrow. | |
250 μg | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Immunohistochemistry (Frozen), Western Blot | |
OX-82 | |
Unconjugated | |
Mouse | |
Protein G | |
RUO | |
Rat | |
Antibody | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction