Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Myeloid Specific Antigen Monoclonal Antibody (BM-2)
GREENER_CHOICE

Catalog No. PIMA533313
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA533313 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA533313 Supplier Invitrogen™ Supplier No. MA533313
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Until recently, immunological markers for myeloid cells have been lacking, especially those which identify different levels of cellular differentiation. The BM series provides a new panel of monoclonal antibodies which stain early precursor and mature forms of human myeloid cells. This panel of monoclonal antibodies reacts with antigenic determinants present in normal myeloid cells and leukemias of similar derivation. BM-2 recognizes a cytoplasmic antigen expressed in mature human granulocytes (polys) residing in lymphoid and non-lymphoid tissues. It does not react with any other cell type in human tissues.

This product provides a new panel of monoclonal antibodies which stain early precursor and mature forms of human myeloid cells. This panel of monoclonal antibodies reacts with antigenic determinants present in normal myeloid cells and leukemias of similar derivation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Myeloid Specific Antigen
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone BM-2
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene MNDA
Gene Accession No. P41218
Gene Alias Ifi204; interferon activated gene 204; MNDA; Myeloid cell nuclear differentiation antigen; PYHIN3; rHin-3; SB135
Gene Symbols MNDA
Host Species Mouse
Immunogen Nuclei from Pokeweed mitogen stimulated human peripheral blood lymphocytes.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4332
Target Species Human, Monkey
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.